Learn More
Alkali Scientific Linsmaier & Skoog Medium, Powder, 1 L
Supplier: Alkali Scientific INS001CP
Linsmaier & Skoog (LS) Medium is a highly effective plant tissue culture medium in powder form, designed to make 1 liter of solution. This medium provides essential nutrients, vitamins, and minerals that promote the growth and differentiation of plant cells in vitro. LS medium is widely used for culturing a variety of plant species, making it a standard tool in agricultural research, genetic studies, and plant biotechnology. It is ideal for use in applications such as callus induction, organogenesis, and somatic embryogenesis. The formulation supports robust plant growth and is often the first choice for many plant tissue culture laboratories.