Learn More
Alkali Scientific Linsmaier & Skoog Medium, Powder, 44g Makes, 10 L
SDP
Catalog No. 5000659672
Encompass_Preferred
Linsmaier & Skoog Medium is a widely used plant tissue culture medium available in powder form, designed to make 10 liters of nutrient solution. The formulation contains essential nutrients, vitamins, and growth regulators required to promote optimal growth and development of plant tissues in vitro. It is commonly used for cultivating both herbaceous and woody plants, and is ideal for applications such as callus induction, root and shoot development, and somatic embryogenesis. LS Medium is a cornerstone of plant tissue culture research and is frequently used in agricultural studies, plant propagation, and genetic modification experiments.
Catalog No. 50-006-59672 Supplier Alkali Scientific Supplier No. INS002CP
Linsmaier & Skoog Medium is a widely used plant tissue culture medium available in powder form, designed to make 10 liters of nutrient solution. The formulation contains essential nutrients, vitamins, and growth regulators required to promote optimal growth and development of plant tissues in vitro. It is commonly used for cultivating both herbaceous and woody plants, and is ideal for applications such as callus induction, root and shoot development, and somatic embryogenesis. LS Medium is a cornerstone of plant tissue culture research and is frequently used in agricultural studies, plant propagation, and genetic modification experiments.