Learn More
Bioworld Linsmaier and Skoog Medium, 25 L
Catalog No. 50254641
Encompass_Preferred
Linsmaier & Skoog MediumContains the Macronutrients, Micronutrients and Vitamins as described by Linsmaier & Skoog (1965) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.43 g/LpH: 3.5–4.5Soluble in water Medium may be sold as Murashige & Skoog (MS) Medium with Minimal Organics (MSMO) by some media suppliers
Catalog No. 50-254-641 Supplier Bioworld Supplier No. 306300452
Linsmaier & Skoog MediumContains the Macronutrients, Micronutrients and Vitamins as described by Linsmaier & Skoog (1965) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.43 g/LpH: 3.5–4.5Soluble in water Medium may be sold as Murashige & Skoog (MS) Medium with Minimal Organics (MSMO) by some media suppliers