Learn More
Bioworld Linsmaier and Skoog Vitamin Concentration (Syringe Vials), 100 mL
Catalog No. 50254643
Encompass_Preferred
Linsmaier & Skoog Vitamin ConcentrationContains the Vitamins as described by Linsmaier & Skoog (1965)Package includes 10 prepared, filtered, sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol 100.00 mg/L Thiamine HCl 0.40 mg/L Total: 4402.49 mg/L
Catalog No. 50-254-643 Supplier Bioworld Supplier No. 306300461
Linsmaier & Skoog Vitamin ConcentrationContains the Vitamins as described by Linsmaier & Skoog (1965)Package includes 10 prepared, filtered, sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol 100.00 mg/L Thiamine HCl 0.40 mg/L Total: 4402.49 mg/L