Learn More
Bioworld Linsmaier and Skoog Vitamin Mix, 100 mL
Catalog No. 50254644
Encompass_Preferred
Linsmaier & Skoog Vitamin MixContains the Vitamins as described by Linsmaier & Skoog (1965)Package contains 10.04 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol 100.00 mg/L Thiamine HCl 0.40 mg/L
Catalog No. 50-254-644 Supplier Bioworld Supplier No. 306300471
Linsmaier & Skoog Vitamin MixContains the Vitamins as described by Linsmaier & Skoog (1965)Package contains 10.04 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol 100.00 mg/L Thiamine HCl 0.40 mg/L