Learn More
Electron Microscopy Sciences Lint-Free Cloth, 13 x 13"
SDP
Catalog No. 50949547
Encompass_Preferred
A twill jean cotton wipe made from 100% cotton fiber that is woven in an extremely tight construction. The extractable trace elements are effectively reduced in the cleaning process which removes impurities in the cotton as well as any starches or binders used in the manufacturing process.
Size: 13 x 13"
This item is not returnable. View return policy
Catalog No. 50-949-547 Supplier Electron Microscopy Sciences Supplier No. 71753
A twill jean cotton wipe made from 100% cotton fiber that is woven in an extremely tight construction. The extractable trace elements are effectively reduced in the cleaning process which removes impurities in the cotton as well as any starches or binders used in the manufacturing process.
Size: 13 x 13"