Learn More
Electron Microscopy Sciences Lint-Free Cloth, 9 x 9"
SDP
Supplier:  Electron Microscopy Sciences 71750
Encompass_Preferred
A twill jean cotton wipe made from 100% cotton fiber that is woven in an extremely tight construction. The extractable trace elements are effectively reduced in the cleaning process which removes impurities in the cotton as well as any starches or binders used in the manufacturing process.
Size: 9 x 9"
Catalog No. 50-949-325