Learn More
Medchemexpress LLC HY-101764 10mg Medchemexpress, Lintitript CAS:136381-85-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10176410MG
Encompass_Preferred
Medchemexpress, HY-101764 10mg Lintitript CAS:136381-85-6 Lintitript (SR 27897) is a highly potent, selective, orally active, competitive and non-peptide cholecystokinin (CCK1) receptor antagonist with an EC50 of 6 nM and a Ki of 0.2 nM. Lintitript displays > 33-fold selectivity more selective for CCK1 than CCK2 receptors (EC50 value of 200 nM). Lintitript increases plasma concentration of leptin and food intake as well as plasma concentration of insulin[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7867