Learn More
TARGETMOL CHEMICALS INC LINTITRIPT 5MG
SDP
Catalog No. 502581205
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lintitript (SR 27897) is a selective antagonist of CCK1 with EC50s of 6 nM and 200 nM for CCK1 and CCK2. The Ki value is 0.2 nM for CCK1. purity: 98%
Catalog No. 50-258-1205 Supplier TARGETMOL CHEMICALS INC Supplier No. T157595MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lintitript (SR 27897) is a selective antagonist of CCK1 with EC50s of 6 nM and 200 nM for CCK1 and CCK2. The Ki value is 0.2 nM for CCK1. purity: 98%