Learn More
Medchemexpress LLC Linuron-d6 | 1219804-76-8 | 99.8% | 1 MG
SDP
Catalog No. 5000410818
Encompass_Preferred
Linuron-d6 is the deuterium labeled Linuron (HY-B1866). Linuron is a phenylurea herbicide used to control grass and weeds in agriculture and orchards. It acts as a photosystem II inhibitor and a competitive androgen receptor (AR) antagonist with a Ki of 100 μM. Linuron exhibits reproductive toxicity in animals as an endocrine disruptor. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a photosystem II inhibitor
  • Functions as a competitive androgen receptor (AR) antagonist
  • Suitable for research use only
Catalog No. 50-004-10818 Supplier Medchemexpress LLC Supplier No. HYB1866S1MG
Linuron-d6 is the deuterium labeled Linuron (HY-B1866). Linuron is a phenylurea herbicide used to control grass and weeds in agriculture and orchards. It acts as a photosystem II inhibitor and a competitive androgen receptor (AR) antagonist with a Ki of 100 μM. Linuron exhibits reproductive toxicity in animals as an endocrine disruptor. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a photosystem II inhibitor
  • Functions as a competitive androgen receptor (AR) antagonist
  • Suitable for research use only