Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linuron-d6 | 1219804-76-8 | 99.8% | 255.13 g/mol | C9H4D6Cl2N2O2 | 10 MG
SDP

Catalog No. 5000297172
Encompass_Preferred

Linuron-d6 is the deuterium-labeled isotopologue of the phenylurea herbicide linuron, provided as a high-purity analytical standard for use as an internal standard or tracer in quantitative NMR, GC-MS, and LC-MS analyses.

  • High purity (99.8%).
  • Molecular formula C9H4D6Cl2N2O2 and molecular weight 255.13 g/mol.
  • Intended for use as an internal standard or tracer in analytical workflows.
  • Available in small pack sizes suitable for reference standards.

Catalog No. 50-002-97172 Supplier Medchemexpress LLC Supplier No. HYB1866S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linuron-d6 is the deuterium-labeled isotopologue of the phenylurea herbicide linuron, provided as a high-purity analytical standard for use as an internal standard or tracer in quantitative NMR, GC-MS, and LC-MS analyses.

  • High purity (99.8%).
  • Molecular formula C9H4D6Cl2N2O2 and molecular weight 255.13 g/mol.
  • Intended for use as an internal standard or tracer in analytical workflows.
  • Available in small pack sizes suitable for reference standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.