Learn More
Medchemexpress LLC Linzagolix | 935283-04-8 | 99.68% | 508.42 | 100 MG
SDP
Catalog No. 5000345434
Encompass_Preferred
Linzagolix is a potent, non-peptide, and orally active GnRH antagonist. It can be used for research related to uterine fibroids, endometriosis, and adenomyosis.
  • Potent, non-peptide, and orally active GnRH antagonist.
  • Can be used for uterine fibroids, endometriosis, and adenomyosis research.
  • Reduces cyst volume in a rat model of endometriosis.
  • Suppresses hypothalamic-pituitary-gonadal function by reducing serum luteinizing hormone and testosterone levels.
Catalog No. 50-003-45434 Supplier Medchemexpress LLC Supplier No. HY10909310MG
Linzagolix is a potent, non-peptide, and orally active GnRH antagonist. It can be used for research related to uterine fibroids, endometriosis, and adenomyosis.
  • Potent, non-peptide, and orally active GnRH antagonist.
  • Can be used for uterine fibroids, endometriosis, and adenomyosis research.
  • Reduces cyst volume in a rat model of endometriosis.
  • Suppresses hypothalamic-pituitary-gonadal function by reducing serum luteinizing hormone and testosterone levels.