Learn More
Medchemexpress LLC Linzagolix | 935283-04-8 | 99.7% | 508.4 g/mol | C22H15F3N2O7S | 5 MG
SDP
Catalog No. 5000263441
Encompass_Preferred
Linzagolix is a non-peptide, orally active gonadotropin-releasing hormone (GnRH) receptor antagonist used in laboratory research into estrogen-dependent conditions such as uterine fibroids, endometriosis, and adenomyosis. Supplied as a small, high-purity solid for research use only (not for human or veterinary use).
  • Non-peptide, orally active GnRH antagonist
  • Applicable to uterine fibroids, endometriosis, and adenomyosis research
  • High chemical purity suitable for analytical and pharmacological studies
  • Supplied in a small 5 mg pack for assay development and pilot studies
  • Molecular weight 508.4 g/mol
Catalog No. 50-002-63441 Supplier Medchemexpress LLC Supplier No. HY1090935MG
Linzagolix is a non-peptide, orally active gonadotropin-releasing hormone (GnRH) receptor antagonist used in laboratory research into estrogen-dependent conditions such as uterine fibroids, endometriosis, and adenomyosis. Supplied as a small, high-purity solid for research use only (not for human or veterinary use).
  • Non-peptide, orally active GnRH antagonist
  • Applicable to uterine fibroids, endometriosis, and adenomyosis research
  • High chemical purity suitable for analytical and pharmacological studies
  • Supplied in a small 5 mg pack for assay development and pilot studies
  • Molecular weight 508.4 g/mol