Learn More
Medchemexpress LLC Linzagolix | 935283-04-8 | 99.7% | 508.42 | 100 MG
SDP
Catalog No. 5000344288
Encompass_Preferred
Linzagolix is a potent, non-peptide, and orally active GnRH antagonist. It can be used for uterine fibroids, endometriosis, and adenomyosis research. For research use only, not for sale to patients.
  • Potent GnRH antagonist
  • Non-peptide compound
  • Orally active
  • Used for uterine fibroids research
  • Used for endometriosis research
  • Used for adenomyosis research
Catalog No. 50-003-44288 Supplier Medchemexpress LLC Supplier No. HY10909350MG
Linzagolix is a potent, non-peptide, and orally active GnRH antagonist. It can be used for uterine fibroids, endometriosis, and adenomyosis research. For research use only, not for sale to patients.
  • Potent GnRH antagonist
  • Non-peptide compound
  • Orally active
  • Used for uterine fibroids research
  • Used for endometriosis research
  • Used for adenomyosis research