Learn More
Medchemexpress LLC Linzagolix | 935283-04-8 | 99.7% | 508.42 | 50 MG
SDP
Catalog No. 5000344156
Encompass_Preferred
Linzagolix is a potent, non-peptide, and orally active GnRH antagonist. It can be used for uterine fibroids, endometriosis, and adenomyosis research. For research use only.
  • Potent, non-peptide, and orally active GnRH antagonist
  • Used for uterine fibroids, endometriosis, and adenomyosis research
Catalog No. 50-003-44156 Supplier Medchemexpress LLC Supplier No. HY109093100MG
Linzagolix is a potent, non-peptide, and orally active GnRH antagonist. It can be used for uterine fibroids, endometriosis, and adenomyosis research. For research use only.
  • Potent, non-peptide, and orally active GnRH antagonist
  • Used for uterine fibroids, endometriosis, and adenomyosis research