Learn More
ILEX LIFE SCIENCES LLC LIONEX EndoTrap HD 1 ml resin FPLC column Endotoxin Removal FPLC Column 1 ML 1/EA
SDP
Catalog No. 5000580377
Encompass_Preferred
EndoTrap HD FPLC Column 1 ml - 8 x 35 mm 1 ml endotoxin (LPS) removal resin in a fast protein liquid chromatography (FPLC) column EndoTrap HD endotoxin removal products manufactured in Germany by LIONEX are intended for in vitro quantitative removal of lipopolysaccharide (LPS) from biological samples in aqueous solutions such as proteins antibodies cell extracts phage and nucleic acids EndoTrap HD can be used in small scale processes for R D and large scale manufacturing processes It can be applied in early or late biomanufacturing process steps EndoTrap HD is based on hydrophilic dimensionally stable affinity matrix with excellent pressure/flow characteristics LPS Binding Capacity 5 000 000 EU/mL LPS Removal Rate 99 99% Sample Recovery Rate 95% pH Sample Range 4-10 Non-toxic no cytotoxic effects Re-use more than 10 times Suitable for high-salt conditions
Catalog No. 50-005-80377 Supplier ILEX LIFE SCIENCES LLC Supplier No. LET0068
EndoTrap HD FPLC Column 1 ml - 8 x 35 mm 1 ml endotoxin (LPS) removal resin in a fast protein liquid chromatography (FPLC) column EndoTrap HD endotoxin removal products manufactured in Germany by LIONEX are intended for in vitro quantitative removal of lipopolysaccharide (LPS) from biological samples in aqueous solutions such as proteins antibodies cell extracts phage and nucleic acids EndoTrap HD can be used in small scale processes for R D and large scale manufacturing processes It can be applied in early or late biomanufacturing process steps EndoTrap HD is based on hydrophilic dimensionally stable affinity matrix with excellent pressure/flow characteristics LPS Binding Capacity 5 000 000 EU/mL LPS Removal Rate 99 99% Sample Recovery Rate 95% pH Sample Range 4-10 Non-toxic no cytotoxic effects Re-use more than 10 times Suitable for high-salt conditions