Learn More
Medchemexpress LLC Liothyronine-13C6-1 | 1213431-76-5 | 98.1% | C913C6H12I3NO4 | 100 UG
SDP
Catalog No. 5000432110
Encompass_Preferred
Liothyronine-13C6-1 is a 13C-labeled Liothyronine, a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32110 Supplier Medchemexpress LLC Supplier No. HYA0070AS2100UG
Liothyronine-13C6-1 is a 13C-labeled Liothyronine, a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS