Learn More
Med Vet International Lip Retractor, 6in, J1056, Each
Catalog No. 5000585619
Encompass_Preferred
The lip or cheek retractor gently and atraumatically pulls the lips back for clear access to the teeth. The tips of the instruments are coated in plastisol to minimize discomfort. It can be adjusted by slightly bending it, 6 1/2in. long and autoclavable.
Catalog No. 50-005-85619 Supplier Med Vet International Supplier No. JOR1056
The lip or cheek retractor gently and atraumatically pulls the lips back for clear access to the teeth. The tips of the instruments are coated in plastisol to minimize discomfort. It can be adjusted by slightly bending it, 6 1/2in. long and autoclavable.