Learn More
Aviva Systems Biology Corp LIPA Antibody - N-terminal region (ARP61109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIPA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human LIPA. Alternate names: CESD, LALApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit, Rat, Sheep
Catalog No. 50-287-256