Learn More
Cayman Chemical LIpase ActIvIty asy KIt 96Wel
SDP
Catalog No. 502893325
Encompass_Preferred
Lipases perform essential roles in the digestion, transportation, and processing of dietary lipids by controlling the clearance of triglyceride-rich lipoproteins from the circulation. Manipulating lipolysis has therapeutic potential in the metabolic disorders frequently associated with obesity. Cayman’s Lipase Activity Assay provides a fluorescence-based method for detecting lipase activity in plasma, serum, tissue homogenates, and cell culture samples. In the assay, lipase hydrolyzes arachidonoyl-1-thioglycerol to arachidonic acid and thioglycerol. Thioglycerol reacts with the thiol fluorometric detector to yield a highly fluorescent product which can be analyzed with an excitation wavelength of 380-390 nm and an emission wavelength of 510-520 nm.
Catalog No. 50-289-3325 Supplier Cayman Chemical Supplier No. 70064096 WELL
Lipases perform essential roles in the digestion, transportation, and processing of dietary lipids by controlling the clearance of triglyceride-rich lipoproteins from the circulation. Manipulating lipolysis has therapeutic potential in the metabolic disorders frequently associated with obesity. Cayman’s Lipase Activity Assay provides a fluorescence-based method for detecting lipase activity in plasma, serum, tissue homogenates, and cell culture samples. In the assay, lipase hydrolyzes arachidonoyl-1-thioglycerol to arachidonic acid and thioglycerol. Thioglycerol reacts with the thiol fluorometric detector to yield a highly fluorescent product which can be analyzed with an excitation wavelength of 380-390 nm and an emission wavelength of 510-520 nm.