Learn More
Crystal Chem Inc Lipase Assay Kitv
SDP
Catalog No. 501947674
Encompass_Preferred
The Lipase Assay is a highly sensitive assay that is used to quantify levels of human lipase in serum and plasma. Highlights: Wide linear range of 0 - 250 U/L. Liquid stable format requires no reagent preparation and reduces waste. Results in as little as ten minutes. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7674 Supplier Crystal Chem Inc Supplier No. 80209
The Lipase Assay is a highly sensitive assay that is used to quantify levels of human lipase in serum and plasma. Highlights: Wide linear range of 0 - 250 U/L. Liquid stable format requires no reagent preparation and reduces waste. Results in as little as ten minutes. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.