Learn More
Medchemexpress LLC Lipase from Candida antarctica (immobilized) | 9001-62-1 | 10g
SDP
Catalog No. 5000267112
Encompass_Preferred
Lipase Candida antarctica (Immobilized) is an immobilized lipase isolated from Candida antarctica fraction B Lipase Candida antarctica (Immobilized) features high stability and selectivity Lipase Candida antarctica (Immobilized) is widely used in fields such as chemical synthesis food oil modification and biodiesel production[1][2]
Catalog No. 50-002-67112 Supplier Medchemexpress LLC Supplier No. HYY1422D10G
Add to Cart
Add to Cart
Lipase Candida antarctica (Immobilized) is an immobilized lipase isolated from Candida antarctica fraction B Lipase Candida antarctica (Immobilized) features high stability and selectivity Lipase Candida antarctica (Immobilized) is widely used in fields such as chemical synthesis food oil modification and biodiesel production[1][2]