Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipase from human pancreas
Catalog No. 502908155
Encompass
Pancreatic lipase fulfills a key function in dietary fat absorption by hydrolyzing triglycerides into diglycerides and subsequently into monoglycerides and free fatty acids.
Catalog No. 50-290-8155 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRE0028100UN
Pancreatic lipase fulfills a key function in dietary fat absorption by hydrolyzing triglycerides into diglycerides and subsequently into monoglycerides and free fatty acids.