Learn More
Medchemexpress LLC Lipase from Pseudomonas fluorescens (immobilized) | 9001-62-1 | MFCD00131509 | 1g
SDP
Catalog No. 5000267111
Encompass_Preferred
Lipase Pseudomonas fluorescens (Immobilized) comes from Pseudomonas fluorescens and shows good applicability in ester synthesis of food industry the biofuel synthesis the synthesis of enantiomeric pure chiral drugs and so on[1]
Catalog No. 50-002-67111 Supplier Medchemexpress LLC Supplier No. HYY1422I1G
Lipase Pseudomonas fluorescens (Immobilized) comes from Pseudomonas fluorescens and shows good applicability in ester synthesis of food industry the biofuel synthesis the synthesis of enantiomeric pure chiral drugs and so on[1]