Learn More
Medchemexpress LLC Lipase, Pseudomonas fluorescens (immobilized) | 9001-62-1 | MFCD00131509 | 500mg
SDP
Catalog No. 5000267995
Encompass_Preferred
Lipase Pseudomonas fluorescens (Immobilized) comes from Pseudomonas fluorescens and shows good applicability in ester synthesis of food industry the biofuel synthesis the synthesis of enantiomeric pure chiral drugs and so on[1]
Catalog No. 50-002-67995 Supplier Medchemexpress LLC Supplier No. HYY1422I500MG
Lipase Pseudomonas fluorescens (Immobilized) comes from Pseudomonas fluorescens and shows good applicability in ester synthesis of food industry the biofuel synthesis the synthesis of enantiomeric pure chiral drugs and so on[1]