Learn More
Diazyme Laboratories Lipase Test Kit, dual vial, liquid, 300 tests, 2 x 50 mL R1, 2 x 11 mL R2, 12 months stability, refrigerated storage, enzymatic test
Catalog No. NC2754174
Encompass_Preferred
Lipase Test Kit, dual vial, liquid, 300 tests, 2 x 50 mL R1, 2 x 11 mL R2, 12 months stability, refrigerated storage, enzymatic test
Catalog No. NC2754174 Supplier Diazyme Laboratories Supplier No. DZ132AK
Lipase Test Kit, dual vial, liquid, 300 tests, 2 x 50 mL R1, 2 x 11 mL R2, 12 months stability, refrigerated storage, enzymatic test