Learn More
ABclonal Technology LIPH Rabbit pAb
SDP
Supplier:  ABclonal Technology A152151000UL
Encompass_Preferred
This gene encodes a membrane-bound member of the mammalian triglyceride lipase family. It catalyzes the production of 2-acyl lysophosphatidic acid (LPA), which is a lipid mediator with diverse biological properties that include platelet aggregation, smooth muscle contraction, and stimulation of cell proliferation and motility.
Catalog No. 50-001-09236