Learn More
Aviva Systems Biology Corp LIPI Antibody - middle region (ARP45940_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45940_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIPI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LIPI. Alternate names: LPDL, PRED5, CT17ApplicationWB. Species reactivity: Human, Pig, Rabbit
Catalog No. 50-014-53910