Learn More
Medchemexpress LLC HY-138171 25mg Medchemexpress, Lipid 5 CAS:2089251-33-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13817125MG
Encompass_Preferred
Medchemexpress, HY-138171 25mg Lipid 5 CAS:2089251-33-0 Lipid 5 is an amino lipid that affords efficient mRNA delivery in rodent and primate models. Lipid 5 shows optimal pharmacokinetics and non-toxic side effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1518