Learn More
Biotium LipidSpot™ 610 Lipid Droplet Stain
Catalog No. NC1609536
Encompass_Preferred
LipidSpot™ 610 dyes rapidly stains lipid droplets with red/far-red fluorescence for detection in the Texas Red or Cy5 channels. For live or fixed cells, no wash step required. Note: due to its broad emission, we do not recommend pairing LipidSpot™ 610 with other red or far-red probes.
Catalog No. NC1609536 Supplier Biotium Supplier No. 70069T
LipidSpot™ 610 dyes rapidly stains lipid droplets with red/far-red fluorescence for detection in the Texas Red or Cy5 channels. For live or fixed cells, no wash step required. Note: due to its broad emission, we do not recommend pairing LipidSpot™ 610 with other red or far-red probes.