Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipid A diphosphoryl from5MG
Catalog No. 502914420
Encompass
Lipid A is the fundamental structure of lipopolysaccharides (LPS) present in bacteria.
Catalog No. 50-291-4420 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L53995MG
Lipid A is the fundamental structure of lipopolysaccharides (LPS) present in bacteria.