Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipid A monophosphoryl Sal
Catalog No. 502905917
Encompass
Lipid A is one of the components of the lipopolysaccharide molecule of Gram-negative bacteria. It is responsible for the toxicity of these bacteria. It has two glucosamine units with a (1--6) glycosidic linkage.
Catalog No. 50-290-5917 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SBR000122ML
Lipid A is one of the components of the lipopolysaccharide molecule of Gram-negative bacteria. It is responsible for the toxicity of these bacteria. It has two glucosamine units with a (1--6) glycosidic linkage.