Learn More
Cayman Chemical LIpId Catechol 1mg
SDP
Catalog No. 502919095
Encompass_Preferred
A lipid used in the formation of lipid prodrug nanoassemblies; lipid catechol-CIP-PBA LPNAs inhibit the formation of, and disrupt preformed, S. aureus biofilms and eradicate staphylococci in a mouse model of peritoneal S. aureus infection; lipid catechol-BTZ LPNAs reduce tumor growth and increase survival in a 4T1 murine mammary carcinoma model
Catalog No. 50-291-9095 Supplier Cayman Chemical Supplier No. 386651 MG
A lipid used in the formation of lipid prodrug nanoassemblies; lipid catechol-CIP-PBA LPNAs inhibit the formation of, and disrupt preformed, S. aureus biofilms and eradicate staphylococci in a mouse model of peritoneal S. aureus infection; lipid catechol-BTZ LPNAs reduce tumor growth and increase survival in a 4T1 murine mammary carcinoma model