Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys TruLab Lipid Level 2 is a lyophilized 'pathologic' high lipid multi-control (3x3mL)
Catalog No. NC2874336
Encompass_Preferred
DiaSys TruLab Lipid Level 2 is a lyophilized 'pathologic' high control based on human blood material (serum) with additives of purified material of human origin. For veterinary or research use only. Contains Apolipoprotein A1, Apolipoprotein B, Cholesterol, HDL, LDL, NEFA, Phospholipids, Triglycerides, Lp-PLA2. DiaSys TruLab Lipid Level 2 must be stored refrigerated. Once reconstituted, DiaSys TruLab Lipid Level 2 is stable for 30 days when kept frozen.
Catalog No. NC2874336 Supplier DIASYS DIAGNOSTIC SYSTEMS USA LLC Supplier No. 590309911065
DiaSys TruLab Lipid Level 2 is a lyophilized 'pathologic' high control based on human blood material (serum) with additives of purified material of human origin. For veterinary or research use only. Contains Apolipoprotein A1, Apolipoprotein B, Cholesterol, HDL, LDL, NEFA, Phospholipids, Triglycerides, Lp-PLA2. DiaSys TruLab Lipid Level 2 must be stored refrigerated. Once reconstituted, DiaSys TruLab Lipid Level 2 is stable for 30 days when kept frozen.