Learn More
Omega Scientific Inc Lipid Depleted FBS, 500 mL
Catalog No. NC2117606
Encompass_Preferred
Lipid Depleted Fetal Bovine Serum is produced by using fumed silica precipitation method for removing lipids. The fumed silica powder is mixed into the serum and centrifuged. The lipids attach to the silica in the pellet and the supernatant contains only the serum. After treatment the level of cholesterol is < 10mg/100ml. Lipid Depleted FBS is suitable for lipid-metabolism studies or Arteriosclerosis research. This product is also known as Delipidated FBS. Virus Screened. Mycoplasma Tested. Triple 0.1 Micron Filtered. 500 mL.
This item is not returnable. View return policy
Catalog No. NC2117606 Supplier Omega Scientific Inc Supplier No. FB50
Lipid Depleted Fetal Bovine Serum is produced by using fumed silica precipitation method for removing lipids. The fumed silica powder is mixed into the serum and centrifuged. The lipids attach to the silica in the pellet and the supernatant contains only the serum. After treatment the level of cholesterol is < 10mg/100ml. Lipid Depleted FBS is suitable for lipid-metabolism studies or Arteriosclerosis research. This product is also known as Delipidated FBS. Virus Screened. Mycoplasma Tested. Triple 0.1 Micron Filtered. 500 mL.