Learn More
Alkali Scientific Lipid Depleted Fetal Bovine Se
SDP
Catalog No. 503026811
Encompass_Preferred
CellProLipid Depleted Fetal Bovine Serum is obtained by using fumed silica precipitation method for removing lipids The fumed silica powder is mixed into the serum and centrifuged The lipids attach to the silica in the pellet and the supernatant contains only the serum After treatment the level of cholesterol is 10mg/100ml Lipid Depleted FBS is suitable for lipid-metabolism studies or Arteriosclerosis research This product is also known as Delipidated FBS Lipid-Depleted Fetal Bovine Serum (FBS) is specifically designed for cell culture applications where lipid content may interfere with experimental outcomes By removing lipids this serum reduces variability caused by lipid-mediated signaling pathways and metabolic processes providing a more controlled environment for sensitive assays Ideal for studies in lipid metabolism lipoprotein research and other lipid-sensitive processes this specialized serum supports optimal cell growth while minimizing unwanted lipid-related influenc
Catalog No. 50-302-6811 Supplier Alkali Scientific Supplier No. FB110
CellProLipid Depleted Fetal Bovine Serum is obtained by using fumed silica precipitation method for removing lipids The fumed silica powder is mixed into the serum and centrifuged The lipids attach to the silica in the pellet and the supernatant contains only the serum After treatment the level of cholesterol is 10mg/100ml Lipid Depleted FBS is suitable for lipid-metabolism studies or Arteriosclerosis research This product is also known as Delipidated FBS Lipid-Depleted Fetal Bovine Serum (FBS) is specifically designed for cell culture applications where lipid content may interfere with experimental outcomes By removing lipids this serum reduces variability caused by lipid-mediated signaling pathways and metabolic processes providing a more controlled environment for sensitive assays Ideal for studies in lipid metabolism lipoprotein research and other lipid-sensitive processes this specialized serum supports optimal cell growth while minimizing unwanted lipid-related influenc