Learn More
Cayman Chemical Lipid Droplets Assay Dye Extr
SDP
Catalog No. 502892364
Encompass_Preferred
A Y-secretase modulator selectively inhibits the production of AB42 in CHO cells expressing wild-type APP (IC50 9.8 nM) over Notch cleavage to NICD (IC50 >15.8 UM) decreases brain levels of AB42 in rats at 10 and 40 mg/kg decreases CSF levels of AB40 and AB42 and increases levels of AB38 and AB39 without affecting total AB CSF levels in dogs at 20 mg/kg
Catalog No. 50-289-2364 Supplier Cayman Chemical Supplier No. 60004630 ML
Add to Cart
Add to Cart
A Y-secretase modulator selectively inhibits the production of AB42 in CHO cells expressing wild-type APP (IC50 9.8 nM) over Notch cleavage to NICD (IC50 >15.8 UM) decreases brain levels of AB42 in rats at 10 and 40 mg/kg decreases CSF levels of AB40 and AB42 and increases levels of AB38 and AB39 without affecting total AB CSF levels in dogs at 20 mg/kg