Learn More
Abcam Lipid E x traction Kit (Chloroform Free), 50 Tests
Catalog No. 502215378
Encompass_Preferred
Lipid Extraction Kit (Chloroform-Free) provides a safer, chloroform-free method for efficient lipid isolation from plasma, serum, tissue, and cells. The recovered organic phase can be dried and resuspended for downstream lipid analysis. - Cited in over 15 publications - Supports multiple sample types including - plasma, serum, tissue, suspension cells, and adherent cells. - Chloroform-free - eliminates the use of toxic chloroform in lipid extraction - Efficient lipid isolation – high-yield lipid extraction with minimal contamination - Sufficient for 50 lipid preparations (25 µL sample size)
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5378 Supplier Abcam Supplier No. AB21104450TEST
Lipid Extraction Kit (Chloroform-Free) provides a safer, chloroform-free method for efficient lipid isolation from plasma, serum, tissue, and cells. The recovered organic phase can be dried and resuspended for downstream lipid analysis. - Cited in over 15 publications - Supports multiple sample types including - plasma, serum, tissue, suspension cells, and adherent cells. - Chloroform-free - eliminates the use of toxic chloroform in lipid extraction - Efficient lipid isolation – high-yield lipid extraction with minimal contamination - Sufficient for 50 lipid preparations (25 µL sample size)
The product is subject to the following: Abcam Restricted Use Statement