Learn More
Cayman Chemical LIpId HTO12 50mg
SDP
Catalog No. 502978489
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.432); has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing lipid HTO12 and encapsulating adenine base editor mRNA targeting Pcsk9 reduce hepatic Pcsk9 levels in mice at a reduced ionizable lipid/total lipids-to-mRNA ratio than LNPs containing SM-102
Catalog No. 50-297-8489 Supplier Cayman Chemical Supplier No. 4120250 MG
An ionizable cationic lipid (pKa = 6.432); has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing lipid HTO12 and encapsulating adenine base editor mRNA targeting Pcsk9 reduce hepatic Pcsk9 levels in mice at a reduced ionizable lipid/total lipids-to-mRNA ratio than LNPs containing SM-102