Learn More
Kamiya Biomedical LIPID HYDROPEROXIDE KIT LPO-CC
SDP
Catalog No. NC0603338 Shop All Kamiya Biomedical Products
Encompass
LPO-CC (Lipid Peroxides) Kit. 165 mL. For the quantitative colorimetric determination of high levels of lipid peroxides in serum, plasma, and other biological samples. Assay range = 2.0 - 300 nmol/mL. (165 tests with 1 mL test tube, 550 tests with 96-well microtiter plate). Includes calibrator.
Catalog No. NC0603338 Supplier Kamiya Biomedical Supplier No. CC004
LPO-CC (Lipid Peroxides) Kit. 165 mL. For the quantitative colorimetric determination of high levels of lipid peroxides in serum, plasma, and other biological samples. Assay range = 2.0 - 300 nmol/mL. (165 tests with 1 mL test tube, 550 tests with 96-well microtiter plate). Includes calibrator.