Learn More
Cayman Chemical Lipid III-45, Purity: ≥98%, CAS: 2096984-25-5, 50 mg
SDP
Catalog No. 502918383
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of LNPs for the delivery of mRNA in vivo; LNPs containing lipid III-45 and encapsulating a luciferase mRNA reporter accumulate in liver and adipose tissue in mice; LNPs containing lipid III-45 have been used to deliver an mRNA sequence encoding an antibody targeting claudin-18.2 to mice and reduce tumor volume in an MDA-MB-231 mouse xenograft model
Catalog No. 50-291-8383 Supplier Cayman Chemical Supplier No. 3924350 MG
An ionizable cationic lipid; has been used in the generation of LNPs for the delivery of mRNA in vivo; LNPs containing lipid III-45 and encapsulating a luciferase mRNA reporter accumulate in liver and adipose tissue in mice; LNPs containing lipid III-45 have been used to deliver an mRNA sequence encoding an antibody targeting claudin-18.2 to mice and reduce tumor volume in an MDA-MB-231 mouse xenograft model