Learn More
Carbosynth LLC. Lipid IVa (0.1mg vial), 1ea, MW: 1405.7, Molecular formula: C68H130N2O23P2, CAS# 91841-27-9
SDP
Supplier:  Carbosynth LLC. CLP24006S0.1MG
Encompass_Preferred
Application: Synthetic Product Biosynthetic Precursor of Lipopolysaccharide, Storage Temperature: -20°C, References: M. Imoto, S. Kusumoto, T. Shiba, E.Th. Rietschel, C. Galanos, and O. Lüderitz, Tetrahedron Lett., 26, 907 (1985). (Original; Chem. Structure), M. Imoto, H. Yoshimura, M. Yamamoto, T. Shimamoto, S. Kusumoto, and T. Shiba, Bull. Chem. Soc. Jpn., 60, 2197 (1987). (Chem. Synthesis)
Catalog No. 50-168-5743