Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipid Mixture 1, Chemically Defined, 100mL
Catalog No. NC1680823
Encompass
liquid, sterile-filtered, BioReagent, Recommended for use in cell culture at 1 to 10 mL per liter of medium.
Catalog No. NC1680823 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L0288100ML
liquid, sterile-filtered, BioReagent, Recommended for use in cell culture at 1 to 10 mL per liter of medium.