Learn More
Cayman Chemical LIpId OA2hydrochlorIde 50mg
SDP
Catalog No. 502917414
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of single-component LNPs for the delivery of siRNA; LNPs containing lipid OA2 and siRNA targeting Socs1 decrease Socs1 expression by ~50% in isolated mouse BMDCs; vaccination with ovalbumin-induced isolated mouse BMDCs transfected with LNPs composed of lipid OA2 and Socs1-targeting siRNA increases survival and decreases tumor volume in a B16-OVA murine melanoma model
Catalog No. 50-291-7414 Supplier Cayman Chemical Supplier No. 3798750 MG
An ionizable cationic lipid; has been used in the generation of single-component LNPs for the delivery of siRNA; LNPs containing lipid OA2 and siRNA targeting Socs1 decrease Socs1 expression by ~50% in isolated mouse BMDCs; vaccination with ovalbumin-induced isolated mouse BMDCs transfected with LNPs composed of lipid OA2 and Socs1-targeting siRNA increases survival and decreases tumor volume in a B16-OVA murine melanoma model