Learn More
Apexbio Technology LLC LIPID PEROX MDA KIT PLUS-100mg
SDP
Supplier:  Apexbio Technology LLC K2312100
Encompass_Preferred
Lipid Peroxidation (MDA) Assay Kit Plus is an essential tool in oxidative stress and lipid peroxidation research It originates from the study of cell membrane lipid damage due to reactive oxygen species (ROS) acting as a marker for oxidative stress The kit measures malondialdehyde (MDA) a by-product of lipid peroxidation using a colorimetric or fluorometric method making it suitable for detecting changes in cellular oxidative states In vitro it exhibits activity typically in the nanomolar to low micromolar range This assay is widely used in applications involving cell models and animal studies to evaluate the impact of potential therapeutic compounds It is also employed in drug screening to assess the antioxidant capacity of new drugs or compounds Experimental concentrations typically depend on the specific research design ensuring flexibility and adaptability across various experimental setups ultimately aiding in the advancement of biomedical research
Catalog No. 50-008-97471