Learn More
Dojindo Molecular Technologies Inc Liperfluo(50UGx5) is used to monitor lipid peroxidation in ferroptosis research, emitting intense fluorescence by a lipid peroxide specific oxidation, and is applicable for microscopy and FCM analysis
SDP
Catalog No. 501903721
Encompass_Preferred
Liperfluo is designed and exclusively developed by Dojindo for detection of lipid peroxides. Among fluorescent probes that detect Reactive Oxygen Species (ROS), Liperfluo is the only compound that can specifically detect lipid peroxides. Since the excitation and emission wavelengths of the oxidized Liperfluo are 524 nm and 535 nm, respectively, both photo-damage and auto-fluorescence can be minimized. The tetraethyleneglycol group linked to one end of the diisoquinoline ring helps its solubility and dispersibility to aqueous buffer. Though Liperfluo's oxidized form is almost nonfluorescent in an aqueous media, it emits fluorescence in lipophilic sites such as cell membranes. Therefore, it can easily be applied to lipid peroxide imaging by a fluorescence microscopy and a flow cytometric analysis for living cells. Liperfluo is widely used in ferroptosis research, a research field Dojindo has a strong portfolio in. (https://www.dojindo.com/ferroptosis-detection-selection-guide/)  
Catalog No. 50-190-3721 Supplier Dojindo Molecular Technologies Inc Supplier No. L24810
Liperfluo is designed and exclusively developed by Dojindo for detection of lipid peroxides. Among fluorescent probes that detect Reactive Oxygen Species (ROS), Liperfluo is the only compound that can specifically detect lipid peroxides. Since the excitation and emission wavelengths of the oxidized Liperfluo are 524 nm and 535 nm, respectively, both photo-damage and auto-fluorescence can be minimized. The tetraethyleneglycol group linked to one end of the diisoquinoline ring helps its solubility and dispersibility to aqueous buffer. Though Liperfluo's oxidized form is almost nonfluorescent in an aqueous media, it emits fluorescence in lipophilic sites such as cell membranes. Therefore, it can easily be applied to lipid peroxide imaging by a fluorescence microscopy and a flow cytometric analysis for living cells. Liperfluo is widely used in ferroptosis research, a research field Dojindo has a strong portfolio in. (https://www.dojindo.com/ferroptosis-detection-selection-guide/)  