Learn More
Cell Biolabs Inc Lipid Quantification Kit (Colorimetric), for unsaturated fatty acids, 100 assays
Supplier:  Cell Biolabs Inc STA613
Encompass_Preferred
Cell Biolabs' Lipid Quantification Kit (Colorimetric) uses a sulfo-phospho-vanillin system to quantify unsaturated fatty acids in a convenient 96-well microplate reader. Supernatants from plasma or serum may be tested directly after centrifugation. Lipids from cells or tissues should be extracted first before quantification. The kit includes sufficient reagents for 100 assays.
Catalog No. 50-673-655