Learn More
Cell Biolabs Inc Lipid Quantification Kit (Fluorometric), for neutral lipds, 100 assays
Catalog No. 501098795
Encompass_Preferred
Cell Biolabs' Lipid Quantification Kit (Fluorometric) quantifies neutral lipids in a convenient 96-well fluorescence-based microplate reader. Supernatants from plasma or serum may be tested directly after centrifugation. Lipids from cells or tissues should be extracted first before quantification. The kit includes sufficient reagents for 100 assays.
Catalog No. 50-109-8795 Supplier Cell Biolabs Inc Supplier No. STA617
Cell Biolabs' Lipid Quantification Kit (Fluorometric) quantifies neutral lipids in a convenient 96-well fluorescence-based microplate reader. Supernatants from plasma or serum may be tested directly after centrifugation. Lipids from cells or tissues should be extracted first before quantification. The kit includes sufficient reagents for 100 assays.