Learn More
Biotium LipidSpot™ 488 Lipid Droplet Stain, 20 uL
Catalog No. NC1669425
Encompass_Preferred
LipidSpot™ Dyes rapidly stains lipid droplets in live cells or fixed cells, with no wash step required. Available with green or red/far-red fluorescence.
Catalog No. NC1669425 Supplier Biotium Supplier No. 70065T
LipidSpot™ Dyes rapidly stains lipid droplets in live cells or fixed cells, with no wash step required. Available with green or red/far-red fluorescence.