Learn More
Cayman Chemical LIpId U-101 10mg
SDP
Catalog No. 502978483
Encompass_Preferred
An ionizable cationic lipid; has been used in the formation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing lipid U-101 and encapsulating mRNA encoding a fusion protein composed of IL-2, a linker, and CD25 inhibit tumor growth in an MC-38 mouse xenograft model
Catalog No. 50-297-8483 Supplier Cayman Chemical Supplier No. 4090210 MG
An ionizable cationic lipid; has been used in the formation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing lipid U-101 and encapsulating mRNA encoding a fusion protein composed of IL-2, a linker, and CD25 inhibit tumor growth in an MC-38 mouse xenograft model